Learn More
JULABO™ External Pt100 Sensor
Product Code. p-8001998
Click to view available options
Length (Metric):
200 mm
300 mm
600 mm
Unit Size:
Each
Product Code. 10135260
Brand: JULABO™ 8981003
Only null left
Add to basket
Add to basket
Stainless steel external Pt100 sensor, for use with a wide variety of JULABO™ water baths, recirculating coolers, and refrigerated and heating circulators.
  • Stainless Steel sensor
  • 1.5 or 3 m cable
  • For use with a wide variety of JULABO™ products:
    • Shaking Water Baths
    • Water Baths
    • Recirculating Coolers
    • Heating Circulators
    • Refrigerated Circulators
Specifications
Description External Pt100 sensor
For Use With (Equipment) JULABO™ Dyneo & Presto
Material Stainless Steel
Size Cable: 1.5 m
Length (Metric) 200 mm
Diameter (Metric) 6 mm