Learn More
exo-2,3-Norbornanedicarboximide, TRC
Specifications
Specifications
| Chemical Name or Material | exo-2,3-Norbornanedicarboximide |
| CAS | 14805-29-9 |
| Molecular Formula | C9 H11 N O2 |
| Quantity | 500 mg |
| Synonym | 4,7-Methano-1H-isoindole-1,3(2H)-dione','hexahydro-','(3aR,4S,7R,7aS)-rel-, 2,3-Norbornanedicarboximide','exo- (8CI), 4,7-Methano-1H-isoindole-1,3(2H)-dione','hexahydro-','(3aα,4β,7β,7aα)-, rel-(3aR,4S,7R,7aS)-Hexahydro-4,7-methano-1H-isoindole-1,3(2H)-dione, NSC 238001, exo-2,3-Norbornanedicarboximide, exo-Norbornane-2,3-dicarboximide, (3aα,4β,7β,7aα)-Hexahydro-4,7-methano-1H-isoindole-1,3(2H)-dione |
| InChI Formula | InChI=1S/C9H11NO2/c11-8-6-4-1-2-5(3-4)7(6)9(12)10-8/h4-7H,1-3H2,(H,10,11,12)/t4-,5+,6+,7- |
| SMILES | O=C1NC(=O)[C@H]2[C@@H]3CC[C@@H](C3)[C@@H]12 |
| Molecular Weight (g/mol) | 165.19 |
| Formula Weight | 165.079 |