Learn More
Justrite Exhaust Fan Lithium Battery Cabinet, ATEX
Click to view available options
Color:
Blue
Unit Size:
Each
Description
- Exhaust fan with filter holding box for high cabinet with 1 or 2 doors
- Outlet diameter: 125 mm (filter excluded)
Specifications
Specifications
| Color | Blue |
| Material | Steel |
| Product Type | Exhaust Fan Cabinet |