Learn More
EX229, MedChemExpress
Product Code. 16480987
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16480987 50 mg 50mg
16474698 10 mg 10mg
16423206 25 mg 25mg
16417417 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16480987 Supplier Medchem Express Supplier No. HY112769
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress EX229, a Benzimidazole derivative, is a potent and allosteric activator of AMP-activated protein kinase (AMPK), with Kds of 0.06 μM, 0.06 μM and 0.51 μM for α1β1γ1, α2β1γ1 and α1β2γ1 in biolayer interferometry, respectively.
EX229, a Benzimidazole derivative, is a potent and allosteric activator of AMP-activated protein kinase (AMPK), with Kds of 0.06 μM, 0.06 μM and 0.51 μM for α1β1γ1, α2β1γ1 and α1β2γ1 in biolayer interferometry, respectively.
Specifications
Chemical Name or Material EX229
CAS 1219739-36-2
Color Pink
Purity Grade Notes Research
Molecular Formula C24H18ClN3O3
Quantity 50 mg
Solubility Information DMSO : 13 mg/mL (30.10 mM; Need ultrasonic and warming)
SMILES O=C(O)C1=CC(OC2=NC3=CC(Cl)=C(C4=CC5=C(N(C)C=C5)C=C4)C=C3N2)=CC=C1C
Molecular Weight (g/mol) 431.87
Formula Weight 431.87
Percent Purity 98.05%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism