Learn More
EVOS™ EVOS™ Onstage Vessel Holder, two Lab-Tek or Lab-Tek-II coverglass chamber slides
Product Code. 15491084
Change view
Click to view available options
Quantity:
1 holder
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15491084 1 holder Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15491084 Supplier EVOS™ Supplier No. AMEPVH069
Only null left
Add to basket
Add to basket
EVOS™ vessel holders and stage plates fit a wide range of vessels, flasks, plates, dishes, and slides.
Product Features:
  • EVOS™ vessel holders allow a perfect fit of your microscope slide, cell culture flask or dish, or micro-well plate to the stage of the EVOS imaging systems for increased precision in sample alignment.
  • EVOS stage plates hold vessel holders, thereby expanding the number of vessel holders available for the EVOS systems.
  • With a snug and secure fit, yet readily switched, these accessories make imaging samples in a range of vessel holders convenient and easy.
TRUSTED_SUSTAINABILITY
Specifications
Type Imaging System Vessel Holder
Product Line EVOS
Description Onstage Vessel Holder, two Lab-Tek or Lab-Tek-II coverglass chamber slides
For Use With (Equipment) EVOS™ FL Auto Imaging System
Holds 2 LabTech-II Chamber Slides, 2 Nunc LabTech-I
Dimensions 159 mm x 109 mm
Quantity 1 holder
Shipping Condition Room Temperature