Learn More
Lenz Laborglasinstrumente™ Evaporating Flask
Especially for use in rotary evaporators. Lenz Laborglasinstrumente™ Evaporating Flasks can be used to make solutions or to hold, contain, collect or measure chemicals and are manufactured from durable DURAN™ glass.
Brand: Lenz Laborglasinstrumente™ B0260551
Additional Details : Weight : 0.44100kg
Description
- With inscription label
- Available in a range of capacities
- Center neck
Specifications
2000 mL |