Learn More
Lenz Laborglasinstrumente™ Evaporating Flask
Especially for use in rotary evaporators. Lenz Laborglasinstrumente™ Evaporating Flasks can be used to make solutions or to hold, contain, collect or measure chemicals and are manufactured from durable DURAN™ glass.
Brand: Lenz Laborglasinstrumente™ B0260471
Additional Details : Weight : 0.05400kg
Description
- With inscription label
- Available in a range of capacities
- Center neck
Specifications
100 mL |