Learn More
Etrasimod, MedChemExpress
Product Code. 16485348
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Product Code. 16485348
Brand: Medchem Express HY12789
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Etrasimod (APD334) is a potent, selective and orally available antagonist of the sphingosine-1-phosphate-1 (S1P1) receptor with an IC50 value of 1.88 nM in CHO cells.
Etrasimod (APD334) is a potent, selective and orally available antagonist of the sphingosine-1-phosphate-1 (S1P1) receptor with an IC50 value of 1.88 nM in CHO cells.
Specifications
Chemical Name or Material Etrasimod
CAS 1206123-37-6
Color White
Purity Grade Notes Research
Molecular Formula C26H26F3NO3
Quantity 10 mg
Synonym APD334
Solubility Information DMSO : ≥ 28 mg/mL (61.20 mM)
SMILES FC(F)(F)C1=CC(COC2=CC=C(NC3=C4CC[C@@H]3CC(O)=O)C4=C2)=CC=C1C5CCCC5
Molecular Weight (g/mol) 457.48
Formula Weight 457.48
Percent Purity 99.81%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation