missing translation for 'onlineSavingsMsg'
Learn More

Etofibrate, MedChemExpress

Product Code. 16463776
Encompass_Preferred
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
This item is not returnable. View return policy

Product Code. 16463776

Brand: Medchem Express HYA0127

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Etofibrate is the ethandiol-1,2 diester of the nicotinic and clofibric acids. Etofibrate has been shown to be a potent hypolipidemic agent in animal and human.

Etofibrate is the ethandiol-1,2 diester of the nicotinic and clofibric acids. Etofibrate has been shown to be a potent hypolipidemic agent in animal and human.

Specifications

Chemical Name or Material Etofibrate
CAS 31637-97-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C18H18ClNO5
Quantity 25 mg
Solubility Information DMSO : ≥ 100 mg/mL (274.88 mM)
SMILES O=C(C1=CC=CN=C1)OCCOC(C(C)(OC2=CC=C(Cl)C=C2)C)=O
Molecular Weight (g/mol) 363.79
Formula Weight 363.79
Percent Purity 99.24%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.