Learn More
Ethynyl Estradiol 17-Beta-D-Glucuronide, TRC
Specifications
Specifications
| Chemical Name or Material | Ethynyl Estradiol 17-beta-D-Glucuronide |
| CAS | 75803-39-3 |
| Molecular Formula | C26 H32 O8 |
| Quantity | 1 mg |
| Synonym | Ethynyl Estradiol 17-B-D-Glucuronide |
| InChI Formula | InChI=1S/C26H32O8/c1-3-26(34-24-21(30)19(28)20(29)22(33-24)23(31)32)11-9-18-17-6-4-13-12-14(27)5-7-15(13)16(17)8-10-25(18,26)2/h1,5,7,12,16-22,24,27-30H,4,6,8-11H2,2H3,(H,31,32)/t16-,17-,18+,19+,20+,21-,22+,24+,25+,26+/m1/s1 |
| SMILES | C[C@]12CC[C@H]3[C@@H](CCc4cc(O)ccc34)[C@@H]1CC[C@@]2(O[C@@H]5O[C@@H]([C@@H](O)[C@H](O)[C@H]5O)C(=O)O)C#C |
| IUPAC Name | (2S,3S,4S,5R,6S)-6-[[(8R,9S,13S,14S,17R)-17-ethynyl-3-hydroxy-13-methyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-yl]oxy]-3,4,5-trihydroxyoxane-2-carboxylic acid |
| Molecular Weight (g/mol) | 472.53 |
| Formula Weight | 472.2097 |