Learn More
Thermo Scientific Alfa Aesar ETHYLENEDIAMINE, 99% 10000ML
Brand:  Thermo Scientific Alfa Aesar A12132.0D
ETHYLENEDIAMINE, 99% 10000ML
Product Code. 11470083
  • ONLINE EXCLUSIVE PRICE
    305.00 €
    301.00 € / 10000mL
    Save 4.00 €
    1% Off