Learn More
Ethyl tridecanoate, 97%
Product Code. 11490768
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11490768 25 g 25g
11480768 5 g 5g
11400778 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11490768 Supplier Thermo Scientific Alfa Aesar Supplier No. B25242.14
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 28267-29-0 | C15H30O2 | 242.403 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 28267-29-0
Molecular Formula C15H30O2
Molecular Weight (g/mol) 242.403
MDL Number MFCD00008978
InChI Key QJYYMNOTJXIOBP-UHFFFAOYSA-N
Synonym tridecanoic acid ethyl ester, tridecanoic acid, ethyl ester, ethyltridecanoate, ethyl=tridecanoate, tridecanoic acid, ethylester, acmc-1cr51, ethyl tridecanoate, n-tridecanoic acid ethyl ester
PubChem CID 119908
IUPAC Name ethyl tridecanoate
SMILES CCCCCCCCCCCCC(=O)OCC
Specifications
Density 0.86
Boiling Point 121°C to 123°C (2.2 mmHg)
Flash Point >110°C (230°F)
Linear Formula CH3(CH2)11CO2CH2CH3
Refractive Index 1.434
Quantity 25 g
Beilstein 1773737
Formula Weight 242.41
Percent Purity 97%
Chemical Name or Material Ethyl tridecanoate