Learn More
Ethyl propionate, 99%
Product Code. 11417987
Change view
Click to view available options
Quantity:
250 mL
1000 mL
Unit Size:
1000mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11417987 1000 mL 1000mL
11427987 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11417987 Supplier Thermo Scientific Alfa Aesar Supplier No. A16650.AU
 Restricted Item
In Stock
Add to basket
CAS: 105-37-3 | C5H10O2 | 102.133 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 105-37-3
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.133
MDL Number MFCD00009308
InChI Key FKRCODPIKNYEAC-UHFFFAOYSA-N
Synonym ethyl propionate, propanoic acid, ethyl ester, propionic ester, propionic ether, propionic acid, ethyl ester, ethylpropionate, propionate d'ethyle, propanoic acid ethyl ester, ethyl n-propionate, propionic acid ethyl ester
PubChem CID 7749
ChEBI CHEBI:41330
IUPAC Name ethyl propanoate
SMILES CCC(=O)OCC
Specifications
Melting Point -74°C
Density 0.891
Boiling Point 99°C
Flash Point 12°C (54°F)
Refractive Index 1.385
Quantity 1000 mL
UN Number UN1195
Beilstein 506287
Merck Index 14,3847
Formula Weight 102.13
Percent Purity 99%
Chemical Name or Material Ethyl propionate