Learn More
Ethyl Oleate-d5, TRC
Specifications
Specifications
| Chemical Name or Material | Ethyl Oleate-d5 |
| CAS | 1217226-00-0 |
| Purity Grade Notes | GC |
| Molecular Formula | C20H33D5O2 |
| Quantity | 10 mg |
| Synonym | (9Z)-9-Octadecenoic Acid Ethyl Ester-d5, Crodamol EO-d5, Esterol 123-d5, Ethyl (Z)-9-Octadecenoate-d5, Ethyl cis-9-Octadecenoate-d5, Ethyl Oleate-d5, Nikkol EOO-d5, Nofable EO 99-d5, Oleic Acid Ethyl Ester |
| InChI Formula | InChI=1S/C20H38O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22-4-2/h11-12H,3-10,13-19H2,1-2H3/b12-11-/i2D3,4D2 |
| SMILES | O=C(OC([2H])([2H])C([2H])([2H])[2H])CCCCCCC/C=C\CCCCCCCC |
| IUPAC Name | 1,1,2,2,2-pentadeuterioethyl (Z)-octadec-9-enoate |
| Molecular Weight (g/mol) | 315.55 |
| Show More |