Learn More
Ethyl methoxyacetate, 98%
Product Code. 11463048
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11463048 50 g 50g
11453048 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11463048 Supplier Thermo Scientific Alfa Aesar Supplier No. A18770.18
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 3938-96-3 | C5H10O3 | 118.132 g/mol
Ethyl methoxyacetate was used as acyl donor during the preparation of enantiomers of several phenylethylamines, as acylation reagent for the aminolysis of 1-phenylethanamine and used in an industrial, lipase-catalyzed kinetic resolution of primary amine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl methoxyacetate was used as acyl donor during the preparation of enantiomers of several phenylethylamines, as acylation reagent for the aminolysis of 1-phenylethanamine and used in an industrial, lipase-catalyzed kinetic resolution of primary amine.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3938-96-3
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.132
MDL Number MFCD00009176
InChI Key JLEKJZUYWFJPMB-UHFFFAOYSA-N
Synonym ethyl methoxyacetate, acetic acid, methoxy-, ethyl ester, 2-methoxy ethyl acetate, ethyl-3-oxabutanoate, methoxyacetic acid ethyl ester, methoxy acetic acid ethyl ester, ethyl-metoxyacetate, methylglycol acetate, ethyl methyloxy acetate, acmc-1cofg
PubChem CID 77544
IUPAC Name ethyl 2-methoxyacetate
SMILES CCOC(=O)COC
Specifications
Density 1
Boiling Point 44°C to 46°C (9 mmHg)
Flash Point 46°C (115°F)
Refractive Index 1.401
Quantity 50 g
UN Number UN3272
Beilstein 1744761
Solubility Information Slightly soluble in water.
Formula Weight 118.13
Percent Purity 98%
Chemical Name or Material Ethyl methoxyacetate