Learn More
Ethyl imidazo[1,2-a]pyridine-2-carboxylate, 95%, Thermo Scientific™
Product Code. 15407517
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15407517 1 g 1g
15417517 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15407517 Supplier Thermo Scientific Alfa Aesar Supplier No. H35384.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 38922-77-9
Molecular Formula C10H10N2O2
Molecular Weight (g/mol) 190.20
MDL Number MFCD02325394
InChI Key GNFACXDTRBVZJE-UHFFFAOYSA-N
Synonym ethyl imidazo 1,2-a pyridine-2-carboxylate, imidazo 1,2-a pyridine-2-carboxylic acid ethyl ester, ethyl h-imidazo 1,2-a pyridine-2-carboxylate, imidazo 1,2-a pyridine-2-carboxylic acid, ethyl ester, imidazo 1,2-a pyridine-2-carboxylicacid, ethyl ester, ethyl 4-hydroimidazo 1,2-a pyridine-2-carboxylate, pubchem17885, acmc-209j2i, ethyl imidazo 1,2-a picolinate, ethylimidazo 1,2-a pyridine-2-carboxylate
PubChem CID 818807
IUPAC Name ethyl imidazo[1,2-a]pyridine-2-carboxylate
SMILES CCOC(=O)C1=CN2C=CC=CC2=N1
Spezifikation
Melting Point 78°C to 80°C
Quantity 1 g
Formula Weight 190.2
Percent Purity 95%
Chemical Name or Material Ethyl imidazo[1,2-a]pyridine-2-carboxylate