Learn More
Ethyl-D5-Amine Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | Ethyl-D5-Amine Hydrochloride |
| CAS | 284474-81-3 |
| Molecular Formula | C2 D5 H2 N . Cl H |
| Quantity | 1 g |
| Synonym | Ethylamine-d5 Hydrochloride, Ethyl-d5-amine HCl |
| InChI Formula | InChI=1S/C2H7N.ClH/c1-2-3;/h2-3H2,1H3;1H/i1D3,2D2; |
| SMILES | Cl.[2H]C([2H])([2H])C([2H])([2H])N |
| IUPAC Name | 1,1,2,2,2-pentadeuterioethanamine;hydrochloride |
| Molecular Weight (g/mol) | 86.58 |
| Formula Weight | 86.0659 |