Learn More
Ethyl Atrarate, TRC
Specifications
Specifications
| Chemical Name or Material | Ethyl Atrarate |
| CAS | 31581-32-5 |
| Molecular Formula | C11H14O4 |
| Quantity | 100 mg |
| Synonym | Atraric Acid Ethyl Ester, Ethyl 3-methylorsellinate, 3,6-Dimethyl-β-resorcylic Acid, Ethyl Ester, Ethyl 2,4-Dihydroxy-3,6-dimethylbenzoate |
| InChI Formula | InChI=1S/C11H14O4/c1-4-15-11(14)9-6(2)5-8(12)7(3)10(9)13/h5,12-13H,4H2,1-3H3 |
| SMILES | CCOC(=O)c1c(C)cc(O)c(C)c1O |
| Molecular Weight (g/mol) | 210.23 |
| Formula Weight | 210.0892 |