Learn More
Ethyl-alpha,alpha-d2-benzene-d5, CDN
Product Code. 30318513
Change view
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30318513 0.1 g 0.1g
30321284 0.25 g 0.25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30318513 Supplier CDN Supplier No. CDND19040.1G
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Ethylbenzene-alpha,alpha,2,3,4,5,6-D7
CAS 84272-90-2
Molecular Formula C8 D7 H3
Quantity 0.1 g
Synonym Benzene-d5, ethyl-1,1-d2- (9 CI, ACI), Ethyl-1,1-d2benzene-d5 (ACI), 1,2,3,4,5-Pentadeuterio-6-(1,1-dideuterioethyl)benzene, Ethyl-α,α-d2-benzene-d5, Pentadeutero(1,1-dideuteroethyl)benzene, Ethylbenzene-d7, Ethylbenzene-α,α,2,3,4,5,6-d7, Ethylbenzene-α,α,2,3,4,5,6-D7
InChI Formula InChI=1 S/C8H10/c1-2-8-6-4-3-5-7-8/h3-7 H,2H2,1H3/i2D2,3 D,4 D,5 D,6 D,7 D
SMILES [2 H]c1c([2 H])c([2 H])c(c([2 H])c1[2 H])C([2 H])([2 H])C
IUPAC Name 1,2,3,4,5-pentadeuterio-6-(1,1-dideuterioethyl)benzene
Molecular Weight (g/mol) 113.2081
Formula Weight 113.1222 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity