Learn More
Ethyl 4-acetyl-5-oxohexanoate, 98%
Product Code. 11358456
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11358456 5 g 5g
11368456 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11358456 Supplier Thermo Scientific Alfa Aesar Supplier No. L17496.06
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 2832-10-2 | C11H21BrO2 | 265.19 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2832-10-2
MDL Number MFCD00009211
Specifications
Density 1.067
Boiling Point 153°C to 154°C (19 mmHg)
Flash Point >110°C (230°F)
Molecular Formula C11H21BrO2
Refractive Index 1.4545
Quantity 5 g
Beilstein 1785189
Synonym undecanoic acid, 11-bromo, 11-bromodecanoic acid, 11-bromo-undecanoic acid, 11-bromoundecanoic acid,tech, 11-bromoundecanoicacid, 11bromoundecanoic acid, 11-bromohendecanoic acid, 11-bromo undecanoic acid, ksc489q1h, 11-bromoundecanoic acid
InChI Key IUDGNRWYNOEIKF-UHFFFAOYSA-N
SMILES C(CCCCCBr)CCCCC(=O)O
IUPAC Name 11-bromoundecanoic acid
Molecular Weight (g/mol) 265.19
PubChem CID 17812
Formula Weight 200.24
Percent Purity 98%
Chemical Name or Material Ethyl 4-acetyl-5-oxohexanoate