Learn More
ethyl(3-methylhexan-2-yl)amine hydrochloride, TRC
Product Code. 30347121
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30347121 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30347121 Supplier Toronto Research Chemicals Supplier No. TRCB43837550MG
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material ethyl(3-methylhexan-2-yl)amine hydrochloride
CAS 1305712-15-5
Molecular Formula C9H21N.HCl
Quantity 50 mg
InChI Formula InChI=1S/C9H21N.ClH/c1-5-7-8(3)9(4)10-6-2;/h8-10H,5-7H2,1-4H3;1H
SMILES Cl.CCCC(C)C(C)NCC
IUPAC Name N-ethyl-3-methylhexan-2-amine;hydrochloride
Molecular Weight (g/mol) 179.73
Formula Weight 179.1441