missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-cyano-3-phenylpyruvate, 97%

Product Code. p-7029484
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11441550 5 g 5g
11451550 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11441550 Supplier Thermo Scientific Alfa Aesar Supplier No. H26951.06
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6362-63-6 | C12H11NO3 | 217.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6362-63-6
Molecular Formula C12H11NO3
Molecular Weight (g/mol) 217.22
MDL Number MFCD00019801
InChI Key YWNRATDDUSMBPR-UHFFFAOYNA-N
Synonym ethyl 3-cyano-3-phenylpyruvate, ethyl phenylcyanopyruvate, cyanophenylpyruvic acid ethyl ester, ethyl cyanophenylpyruvate, ethyl alpha-cyanophenylpyruvate, pyruvic acid, cyanophenyl-, ethyl ester, ethyl .alpha.-cyanophenylpyruvate, wln: ncr x1v1uno2, 4-10-00-03440 beilstein handbook reference, benzenepropanoic acid, .beta.-cyano-.alpha.-oxo-, ethyl ester
PubChem CID 22845
IUPAC Name ethyl 3-cyano-2-oxo-3-phenylpropanoate
SMILES CCOC(=O)C(=O)C(C#N)C1=CC=CC=C1

Specifications

Melting Point 125°C to 129°C
Quantity 5 g
Formula Weight 217.23
Percent Purity 97%
Chemical Name or Material Ethyl 3-cyano-3-phenylpyruvate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.