Learn More
Ethyl 3-Aminopyridine-2-carboxylate, TRC
Specifications
Specifications
| Chemical Name or Material | Ethyl 3-Aminopyridine-2-carboxylate |
| CAS | 27507-15-9 |
| Molecular Formula | C8 H10 N2 O2 |
| Quantity | 100 mg |
| Synonym | 2-Pyridinecarboxylic acid, 3-amino-, ethyl ester, Picolinic acid, 3-amino-, ethyl ester (8CI), 3-Amino-2-(ethoxycarbonyl)pyridine, 3-Aminopyridine-2-carboxylic acid ethyl ester, Ethyl 3-amino-2-picolinate, Ethyl 3-aminopicolinate, Ethyl 3-aminopyridine-2-carboxylate |
| InChI Formula | InChI=1S/C8H10N2O2/c1-2-12-8(11)7-6(9)4-3-5-10-7/h3-5H,2,9H2,1H3 |
| SMILES | CCOC(=O)c1ncccc1N |
| IUPAC Name | ethyl 3-aminopyridine-2-carboxylate |
| Molecular Weight (g/mol) | 166.18 |
| Formula Weight | 166.0742 |