missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-Aminobenzoate Methanesulfonate, TRC

Product Code. 17931585
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17931585 5 g 5g
30136849 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17931585 Supplier Toronto Research Chemicals Supplier No. TRCE6785655G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 886-86-2
Formula Weight 261.0671
InChI Formula InChI=1S/C9H11NO2.CH4O3S/c1-2-12-9(11)7-4-3-5-8(10)6-7;1-5(2,3)4/h3-6H,2,10H2,1H3;1H3,(H,2,3,4)
IUPAC Name ethyl 3-aminobenzoate;methanesulfonic acid
Molecular Formula C9 H11 N O2 . C H4 O3 S
Molecular Weight (g/mol) 261.29
SMILES CCOC(=O)c1cccc(N)c1.CS(=O)(=O)O
Synonym Benzoic acid, 3-amino-, ethyl ester, methanesulfonate (9CI), Benzoic acid, m-amino-, ethyl ester, methanesulfonate (7CI,8CI), Ethyl 3-aminobenzoate methanesulfonate, Ethyl m-aminobenzoate methanesulfonate, Finquel, MS 222, Metacain, Metacaine, NSC 93790, TS 222, Tricaine, Tricaine methanesulfonate
Chemical Name or Material Tricaine Methanesulfonate

Specifications

Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.