Learn More
Ethyl 2-pyridineacetate, 98%
Product Code. 11466876
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11466876 5 g 5g
11476876 25 g 25g
11486876 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11466876 Supplier Thermo Scientific Alfa Aesar Supplier No. B23988.06
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 2739-98-2 | C9H11NO2 | 165.192 g/mol
Ethyl 2-pyridineacetate is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl 2-pyridineacetate is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
Solubility
Soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2739-98-2
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD00006359
InChI Key IUDKTVXSXWAKJO-UHFFFAOYSA-N
Synonym ethyl 2-pyridylacetate, ethyl 2-pyridin-2-yl acetate, ethyl 2-pyridinylacetate, ethyl pyridine-2-acetate, 2-pyridineacetic acid ethyl ester, ethyl 2-pyridineacetate, ethyl 2-2-pyridyl acetate, 2-pyridylacetic acid ethyl ester, ethyl pyridin-2-ylacetate, 2-pyridineacetic acid, ethyl ester
PubChem CID 75960
IUPAC Name ethyl 2-pyridin-2-ylacetate
SMILES CCOC(=O)CC1=CC=CC=N1
Specifications
Density 1.084
Boiling Point 69°C to 70°C (0.05 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.497
Quantity 5 g
Beilstein 123817
Solubility Information Soluble in water.
Formula Weight 165.19
Percent Purity 98%
Chemical Name or Material Ethyl 2-pyridineacetate