missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-nitrobenzoylacetate, 97%

Product Code. p-4089964 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10186397 1 g 1g
10789438 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10186397 Supplier Thermo Scientific Acros Supplier No. 430040010
Add to basket
This item is not returnable. View return policy

CAS: 52119-39-8 | C11H11NO5 | 237.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52119-39-8
Molecular Formula C11H11NO5
Molecular Weight (g/mol) 237.21
MDL Number MFCD03424821
InChI Key OWZNCVIBJQPNEF-UHFFFAOYSA-N
Synonym ethyl 3-2-nitrophenyl-3-oxopropanoate, ethyl 2-nitrobenzoylacetate, acmc-1awhb, ethyl 3-2-nitrophenyl-3-oxo-propanoate, ethyl o-nitrobenzoylacetate, ethyl 2-o-nitrobenzoyl acetate, ethyl 2-2-nitrobenzoyl acetate, ethyl 2-o-nitrobenzoyl-acetate, ethyl 2-nitrobenzoyl acetate
PubChem CID 10561837
IUPAC Name ethyl 3-(2-nitrophenyl)-3-oxopropanoate
SMILES CCOC(=O)CC(=O)C1=CC=CC=C1[N+](=O)[O-]

Specifications

Melting Point 35.0°C to 36.0°C
Color Orange to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 237.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethyl 2-nitrobenzoylacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.