Learn More
Ethyl-2-d1 Alcohol, CDN
Product Code. 30315353
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30315353
Brand: CDN CDND11030.25G
Please to purchase this item. Need a web account? Register with us today!
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Ethanol-2-D
CAS 8-4-6181
Molecular Formula C2 D H5 O
Quantity 0.25 g
Synonym Ethan-2-d-ol (9 CI), Ethyl-2-d alcohol (7 CI, 8 CI), 2-Deuterioethanol, Ethanol-2-D, Ethanol-d1, Ethyl-2-d1 Alcohol
InChI Formula InChI=1 S/C2H6O/c1-2-3/h3H,2H2,1H3/i1D
SMILES [2 H]CCO
IUPAC Name 2-deuterioethanol
Molecular Weight (g/mol) 47.0746
Formula Weight 47.0481 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity