missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-chloro-3-pyridylglyoxylate, 97%

Product Code. 11469270
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11469270 5 g 5g
11459270 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11469270 Supplier Thermo Scientific Alfa Aesar Supplier No. H50153.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 902837-56-3 | C9H8ClNO3 | 213.617 g/mol

Ethyl 2-chloro-3-pyridylglyoxylate is used as a pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2-chloro-3-pyridylglyoxylate is used as a pharmaceutical intermediates.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 902837-56-3
Molecular Formula C9H8ClNO3
Molecular Weight (g/mol) 213.617
MDL Number MFCD07781194
InChI Key ZTIMVJIGNBYGJG-UHFFFAOYSA-N
Synonym ethyl 2-chloro-3-pyridylglyoxylate, ethyl 2-2-chloropyridin-3-yl-2-oxoacetate, ethyl 2-chloropyridin-3-yl oxo acetate, ethyl 2-chloro-3-pyridinyl oxoacetate, 2-chloro-3-pyridylglyoxylic acid ethyl ester, ethyl 2-2-chloro 3-pyridyl-2-oxoacetate, 2-chloro-pyridin-3-yl-oxo-acetic acid ethyl ester
PubChem CID 42553166
IUPAC Name ethyl 2-(2-chloropyridin-3-yl)-2-oxoacetate
SMILES CCOC(=O)C(=O)C1=C(N=CC=C1)Cl

Specifications

Quantity 5 g
Formula Weight 213.62
Percent Purity 97%
Chemical Name or Material Ethyl 2-chloro-3-pyridylglyoxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.