Learn More
Ethyl 2-[3-Fluoro-(trifluoromethyl)phenyl]-4-methyl-thiazole-5-carboxylate, TRC
Specifications
Specifications
| Chemical Name or Material | Ethyl 2-[3-Fluoro-(trifluoromethyl)phenyl]-4-methyl-thiazole |
| CAS | 317319-21-4 |
| Molecular Formula | C14 H11 F4 N O2 S |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C14H11F4NO2S/c1-3-21-13(20)11-7(2)19-12(22-11)8-4-5-9(10(15)6-8)14(16,17)18/h4-6H,3H2,1-2H3 |
| SMILES | CCOC(=O)c1sc(nc1C)c2ccc(c(F)c2)C(F)(F)F |
| IUPAC Name | ethyl 2-[3-fluoro-4-(trifluoromethyl)phenyl]-4-methyl-1,3-thiazole-5-carboxylate |
| Molecular Weight (g/mol) | 333.3 |
| Formula Weight | 333.0447 |