missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2,3-dibromopropionate, 97%

Product Code. 11467437
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11467437 100 g 100g
11457437 25 g 25g
11477437 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11467437 Supplier Thermo Scientific Alfa Aesar Supplier No. A15591.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 3674-13-3 | C5H8Br2O2 | 259.925 g/mol

Ethyl 2,3-dibromopropionate is used as pharmaceutical Intermediates. It is a building block that has been used as a catalyst for the polymerization of ethyl acrylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2,3-dibromopropionate is used as pharmaceutical Intermediates. It is a building block that has been used as a catalyst for the polymerization of ethyl acrylate.

Solubility
Insoluble in water.

Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent, acids, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3674-13-3
Molecular Formula C5H8Br2O2
Molecular Weight (g/mol) 259.925
MDL Number MFCD00000212
InChI Key OENICUBCLXKLJQ-UHFFFAOYSA-N
PubChem CID 97945
IUPAC Name ethyl 2,3-dibromopropanoate
SMILES CCOC(=O)C(CBr)Br

Specifications

Density 1.789
Boiling Point 212°C to 214°C
Flash Point 90°C (194°F)
Odor Irritating
Refractive Index 1.5 00
Quantity 100 g
UN Number UN2922
Beilstein 636046
Synonym ethyl 2,3-dibromopropionate, propanoic acid, 2,3-dibromo-, ethyl ester, ethyl alpha,beta-dibromopropionate, propionic acid, 2,3-dibromo-, ethyl ester, ethyl-2,3-dibromopropionate, 2,3-dibromopropionic acid ethyl ester, ethyl .alpha.,.beta.-dibromopropionate, propionic acid, 2,3-dibromo-, ethyl ester 6ci,7ci,8ci, ethyl2,3-dibromopropionate, acmc-1ahn6
Solubility Information Insoluble in water.
Formula Weight 259.94
Percent Purity 97%
Chemical Name or Material Ethyl 2,3-dibromopropionate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.