Learn More
Ethyl 2,3-Dibromopropanoate, TRC
Product Code. 30375346
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30375346 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30375346 Supplier Toronto Research Chemicals Supplier No. TRCE91584025G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Ethyl 2,3-Dibromopropanoate
CAS 3674-13-3
Molecular Formula C5H8Br2O2
Quantity 25 g
Synonym 2,3-Dibromopropanoic Acid Ethyl Ester, 2,3-Dibromopropionic Acid Ethyl Ester, Ethyl 2,3-Dibromopropionate, Ethyl alpha,beta-Dibromopropionate, NSC 60552
InChI Formula InChI=1S/C5H8Br2O2/c1-2-9-5(8)4(7)3-6/h4H,2-3H2,1H3
SMILES CCOC(=O)C(Br)CBr
IUPAC Name ethyl 2,3-dibromopropanoate
Molecular Weight (g/mol) 259.92
Formula Weight 257.8891