Learn More
Ethyl 2,3,4-Tri-O-benzyl-1-thio-Alpha,Beta-L-fucopyranoside, TRC
Product Code. 30370362
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30370362 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30370362 Supplier Toronto Research Chemicals Supplier No. TRCE927500100MG
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
CAS 169532-17-6
Molecular Formula C29 H34 O4 S
Quantity 100 mg
InChI Formula InChI=1S/C29H34O4S/c1-3-34-29-28(32-21-25-17-11-6-12-18-25)27(31-20-24-15-9-5-10-16-24)26(22(2)33-29)30-19-23-13-7-4-8-14-23/h4-18,22,26-29H,3,19-21H2,1-2H3/t22-,26+,27+,28-,29?/m0/s1
SMILES CCSC1O[C@@H](C)[C@@H](OCc2ccccc2)[C@@H](OCc3ccccc3)[C@@H]1OCc4ccccc4
IUPAC Name (3S,4R,5R,6S)-2-ethylsulfanyl-6-methyl-3,4,5-tris(phenylmethoxy)oxane
Molecular Weight (g/mol) 478.64
Formula Weight 478.2178
Chemical Name or Material Ethyl 2,3,4-Tri-O-benzyl-1-thio-alpha,beta-L-fucopyranoside