Learn More
Ethyl 2-(1-methyl-1H-imidazol-4-yl)acetate, TRC
Specifications
Specifications
| Chemical Name or Material | Ethyl 2-(1-methyl-1H-imidazol-4-yl)acetate |
| CAS | 916792-95-5 |
| Molecular Formula | C8H12N2O2 |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C8H12N2O2/c1-3-12-8(11)4-7-5-10(2)6-9-7/h5-6H,3-4H2,1-2H3 |
| SMILES | CCOC(=O)Cc1cn(C)cn1 |
| IUPAC Name | ethyl 2-(1-methylimidazol-4-yl)acetate |
| Molecular Weight (g/mol) | 168.19 |
| Formula Weight | 168.0899 |