missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1-fluoro-2-oxocyclohexanecarboxylate, 97%, Thermo Scientific Chemicals

Product Code. p-7032771
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15488905 250 mg 250mg
15407317 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15488905 Supplier Thermo Scientific Alfa Aesar Supplier No. H34377.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1578-70-7 | Molecular Formula: C9H13FO3 | Molecular Weight: 188.198 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1578-70-7
Molecular Formula C9H13FO3
Molecular Weight (g/mol) 188.198
MDL Number MFCD01076477
InChI Key WBRNDLRGLOAFCQ-UHFFFAOYSA-N
Synonym ethyl 1-fluoro-2-oxocyclohexanecarboxylate, 1-fluoro-2-oxocyclohexanecarboxylic acid ethyl ester, cyclohexanecarboxylic acid, 1-fluoro-2-oxo-, ethyl ester
PubChem CID 10921276
IUPAC Name ethyl 1-fluoro-2-oxocyclohexane-1-carboxylate
SMILES CCOC(=O)C1(CCCCC1=O)F

Specifications

Boiling Point 66°C (0.35 mmHg)
Quantity 250 mg
Formula Weight 188.2
Percent Purity 97%
Chemical Name or Material Ethyl 1-fluoro-2-oxocyclohexanecarboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.