Learn More
Ethychlozate, TRC
Specifications
Specifications
| Chemical Name or Material | Ethychlozate |
| CAS | 27512-72-7 |
| Molecular Formula | C11 H11 Cl N2 O2 |
| Quantity | 100 mg |
| Synonym | 1H-Indazole-3-acetic acid, 5-chloro-, ethyl ester (8CI, 9CI, ACI), Ethychlozate, Ethyl 5-chloro-1H-indazole-3-acetate, Ethylchlozate, Etychlozate, Figarin, Figaron, IZAA, J 455 |
| InChI Formula | InChI=1S/C11H11ClN2O2/c1-2-16-11(15)6-10-8-5-7(12)3-4-9(8)13-14-10/h3-5H,2,6H2,1H3,(H,13,14) |
| SMILES | CCOC(=O)Cc1n[nH]c2ccc(Cl)cc12 |
| IUPAC Name | ethyl 2-(5-chloro-1H-indazol-3-yl)acetate |
| Molecular Weight (g/mol) | 238.67 |
| Formula Weight | 238.0509 |