Learn More
Ethephon-d4, CDN
Product Code. 30314097
Change view
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30314097 0.01 g 0.01g
30318072 0.005 g 0.01g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30314097 Supplier CDN Supplier No. CDND80550.01G
Please to purchase this item. Need a web account? Register with us today!
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-(Chloroethyl) Phosphonic Acid-d4
CAS 1020719-29-2
Molecular Formula C2 2H4 H2 Cl O3 P
Quantity 0.01 g
Synonym (2-Chloroethyl) Phosphonic Acid-d4, Ethephon D4 (2-Chloroethyl-1,1,2,2-D4), 2-Chloroethanephosphonic-d4 Acid, 2-KhEFK-d4, Amchem-d4, Ethephon-d4, Roll-Fruct-d4, Romtrel-d4, Super Boll-d4, Tomathrel-d4
InChI Formula InChI=1 S/C2H6ClO3P/c3-1-2-7(4,5)6/h1-2H2,(H2,4,5,6)/i1D2,2D2
SMILES [2 H]C([2 H])(Cl)C([2 H])([2 H])P(=O)(O)O
IUPAC Name (2-chloro-1,1,2,2-tetradeuterioethyl)phosphonic acid
Molecular Weight (g/mol) 148.52
Formula Weight 147.9994 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity