missing translation for 'onlineSavingsMsg'
Learn More

Ethamsylate, MedChemExpress

Product Code. 16488658
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16488658

Brand: Medchem Express HYB1074

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Ethamsylate is a haemostatic drug, also inhibits biosynthesis and action of those prostaglandins.

Ethamsylate is a haemostatic drug, also inhibits biosynthesis and action of those prostaglandins.

Specifications

Chemical Name or Material Ethamsylate
CAS 2624-44-4
Color White
Purity Grade Notes Research
Molecular Formula C10H17NO5S
Quantity 10 mg
Synonym Etamsylate
Solubility Information DMSO : 120 mg/mL (455.74 mM; Need ultrasonic) ∣H2O : 50 mg/mL (189.89 mM; Need ultrasonic)
SMILES O=S(C1=CC(O)=CC=C1O)(O)=O.CCNCC
Molecular Weight (g/mol) 263.31
Formula Weight 263.31
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.