Learn More
Estrone Beta-D-Glucuronide Triacetate Methyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | Estrone beta-D-Glucuronide Triacetate Methyl Ester |
| CAS | 27537-72-0 |
| Molecular Formula | C31H38O11 |
| Quantity | 10 mg |
| Synonym | 17-Oxoestra-1,3,5(10)-trien-3-yl-beta-D-glucopyranosiduronic Acid Triacetate Methyl Ester, 3-(beta-D-Glucopyranuronosyloxy)-estra-1,3,5(10)-trien-17-one Methyl Ester Triacetate |
| InChI Formula | InChI=1S/C31H38O11/c1-15(32)38-25-26(39-16(2)33)28(40-17(3)34)30(42-27(25)29(36)37-5)41-19-7-9-20-18(14-19)6-8-22-21(20)12-13-31(4)23(22)10-11-24(31)35/h7,9,14,21-23,25-28,30H,6,8,10-13H2,1-5H3/t21-,22-,23+,25+,26?,27+,28?,30-,31+/m1/s1 |
| SMILES | COC(=O)[C@H]1O[C@@H](Oc2ccc3[C@H]4CC[C@@]5(C)[C@@H](CCC5=O)[C@@H]4CCc3c2)C(OC(=O)C)C(OC(=O)C)[C@@H]1OC(=O)C |
| IUPAC Name | methyl (2S,3S,6S)-3,4,5-triacetyloxy-6-[[(8R,9S,13S,14S)-13-methyl-17-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-3-yl]oxy]oxane-2-carboxylate |
| Molecular Weight (g/mol) | 586.63 |
| Formula Weight | 586.2414 |