Learn More
Estradiol 17Beta-sulfate, TRC
Specifications
Specifications
| Chemical Name or Material | Estradiol 17Beta-sulfate |
| CAS | 3233-69-0 |
| Molecular Formula | C18H24O5S |
| Quantity | 1 mg |
| Synonym | Estra-1,3,5(10)-triene-3,17-diol (17β)-, 17-(hydrogen sulfate) (9CI, ACI), Estradiol, 17-(hydrogen sulfate) (7CI, 8CI), 17β-Estradiol 17-sulfate, 3,17β-Dihydroxyestra-1,3,5(10)-triene 17-sulfate, Estradiol 17-sulfate, Estradiol 17β-sulfate |
| InChI Formula | InChI=1S/C18H24O5S/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)23-24(20,21)22/h3,5,10,14-17,19H,2,4,6-9H2,1H3,(H,20,21,22)/t14-,15-,16+,17+,18+/m1/s1 |
| SMILES | OC1=CC(CC[C@]([C@]2([H])CC[C@@H]3OS(O)(=O)=O)([H])[C@]4([H])CC[C@@]23C)=C4C=C1 |
| Molecular Weight (g/mol) | 352.45 |