missing translation for 'onlineSavingsMsg'
Learn More

Escin (mixture of natural saponins, Technical Grade), TRC

Product Code. 30368818
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30368818 1 g 1g
30372747 100 mg -
30382447 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30368818 Supplier Toronto Research Chemicals Supplier No. TRCE6587901G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6805-41-0
Molecular Formula C55H86O24
Molecular Weight (g/mol) 1131.27
Synonym 3-(4-O-β-D-glucopyranosyl-2-O-β-D-xylopyranosyl-β-D-glucopyranuronoside)escigenin 3-Hydroxy-2-methylbutyrate Acetate, 3,5-Epoxypicene, escin deriv., Oleanane Escin deriv., Aescin, Aescine, Aescusan, Ba 2672, Escusan, Eskuzan, Flebostasin Retard
IUPAC Name (2S,3S,4S,5R)-6-(((3S,4S,6aR,6bS,8R,8aR,9R,10R,14bR)-9-acetoxy-8-hydroxy-4,8a-bis(hydroxymethyl)-4,6a,6b,11,11,14b-hexamethyl-10-(((E)-2-methylbut-2-enoyl)oxy)-1,2,3,4,4a,5,6,6a,6b,7,8,8a,9,10,11,12,12a,14,14a,14b-icosahydropicen-3-yl)oxy)-4-hydroxy-3,5-bis(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-carboxylic acid
SMILES O[C@@H]1C[C@@]2(C)[C@]3(C)CCC4[C@](CO)(C)[C@@H](OC5O[C@H](C(O)=O)[C@@H](O[C@@H]6O[C@H](CO)[C@@H](O)[C@H](O)[C@H]6O)[C@H](O)[C@H]5O[C@@H]7O[C@H](CO)[C@@H](O)[C@H](O)[C@H]7O)CC[C@]4(C)C3CC=C2C8[C@]1(CO)[C@@H](OC(C)=O)[C@H](OC(/C(C)=C/C)=O)C(C)(C)C8

Specifications

Quantity 1 g
InChI Formula InChI=1S/C55H86O24/c1-10-23(2)46(71)79-43-44(72-24(3)60)55(22-59)26(17-50(43,4)5)25-11-12-30-51(6)15-14-32(52(7,21-58)29(51)13-16-53(30,8)54(25,9)18-31(55)61)75-49-41(77-48-38(67)36(65)34(63)28(20-57)74-48)39(68)40(42(78-49)45(69)70)76-47-37(66)35(64)33(62)27(19-56)73-47/h10-11,26-44,47-49,56-59,61-68H,12-22H2,1-9H3,(H,69,70)/b23-10+/t26?,27-,28+,29?,30?,31-,32+,33-,34+,35+,36-,37-,38+,39+,40+,41-,42+,43+,44+,47+,48-,49?,51+,52-,53-,54-,55+/m1/s1
Formula Weight 1130.57
Chemical Name or Material Aescin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.