missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies ErythroClear™ Red Blood Cell Depletion Reagent Kit

Product Code. 17197711
Encompass
Change view
Click to view available options
Quantity:
2 mL cord blood
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17197711 2 mL cord blood 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17197711 Supplier STEMCELL Technologies Supplier No. 01738

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent for processing 2 mL of cord blood

  • ErythroClear™ is intended for the depletion of red blood cells (RBCs) from up to 16 small volume (50 - 100 μL) fresh or frozen cord blood samples at a time
  • ErythroClear™ reagent contains immunomagnetic particles that bind to Glycophorin AB+ cells, which are then selectively depleted using an ErythroClear™ magnet
  • This product is recommended for the depletion of RBCs in cord blood samples prior to downstream assays (e.g. CFU assay, ALDHbr assay)
  • The ErythroClear™ Red Blood Cell Depletion Kit (Catalog #01739) contains an ErythroClear™ magnet and ErythroClear™ reagent
  • The ErythroClear™ magnet is not sold separately and is only available when purchased as a part of this kit
  • The ErythroClear™ Red Blood Cell Depletion Reagent Kit (Catalog #01738) contains ErythroClear™ reagent only, and is suitable as a refill for labs that already have an ErythroClear™ magnet
  • The ErythroClear™ magnet is required for use of the ErythroClear™ reagent

Specifications

Cell Type Hematopoietic Stem and Progenitor Cells, Red Blood Cells
For Use With (Application) Cell Isolation
Quantity 2 mL cord blood
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.