missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies ErythroClear™ Red Blood Cell Depletion Kit

Product Code. 17107721
Encompass
Click to view available options
Quantity:
2 mL cord blood
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 17107721

Brand: STEMCELL Technologies 01739

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Kit including ErythroClear™ magnet and reagent for processing 2 mL of cord blood

  • ErythroClear™ is intended for the depletion of red blood cells (RBCs) from up to 16 small volume (50 - 100 μL) fresh or frozen cord blood samples at a time
  • ErythroClear™ reagent contains immunomagnetic particles that bind to Glycophorin AB+ cells, which are then selectively depleted using an ErythroClear™ magnet
  • This product is recommended for the depletion of RBCs in cord blood samples prior to downstream assays (e.g. CFU assay, ALDHbr assay)
  • The ErythroClear™ Red Blood Cell Depletion Kit (Catalog #01739) contains an ErythroClear™ magnet and ErythroClear™ reagent
  • The ErythroClear™ magnet is not sold separately and is only available when purchased as a part of this kit
  • The ErythroClear™ Red Blood Cell Depletion Reagent Kit (Catalog #01738) contains ErythroClear™ reagent only, and is suitable as a refill for labs that already have an ErythroClear™ magnet
  • The ErythroClear™ magnet is required for use of the ErythroClear™ reagent

Specifications

Cell Type Hematopoietic Stem and Progenitor Cells, Red Blood Cells
For Use With (Application) Cell Isolation
Quantity 2 mL cord blood
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.