Learn More
Ertugliflozin L-pyroglutamic acid, MedChemExpress
Product Code. 16439127
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16439127
Brand: Medchem Express HY15461A
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Ertugliflozin L-pyroglutamic acid (PF-04971729 L-pyroglutamic acid) is a potent, selective and orally active inhibitor of the sodium-dependent glucose cotransporter 2 (SGLT2), with an IC50 of 0.877 nM for h-SGLT2. Has the potential for the treatment of type 2 diabetes mellitus.
Ertugliflozin L-pyroglutamic acid (PF-04971729 L-pyroglutamic acid) is a potent, selective and orally active inhibitor of the sodium-dependent glucose cotransporter 2 (SGLT2), with an IC50 of 0.877 nM for h-SGLT2. Has the potential for the treatment of type 2 diabetes mellitus.
Specifications
Chemical Name or Material Ertugliflozin L-pyroglutamic acid
CAS 1210344-83-4
Color White
Purity Grade Notes Research
Molecular Formula C27H32ClNO10
Quantity 5 mg
Synonym PF-04971729 L-pyroglutamic acid
Solubility Information DMSO : ≥ 125 mg/mL (220.85 mM)
SMILES ClC1=CC=C([C@]23O[C@@](CO)(CO3)[C@@H](O)[C@H](O)[C@H]2O)C=C1CC4=CC=C(OCC)C=C4.O=C5CC[C@@H](C(O)=O)N5
Molecular Weight (g/mol) 566.0
Formula Weight 566.0
Percent Purity 99.77%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism