Learn More
OHAUS™ Erlenmeyer Flask Clamp
Shop All Ohaus Products
Click to view available options
For Use With (Equipment):
1 L Erlenmeyer Flask
2 L Erlenmeyer Flask
2.8 L Erlenmeyer Flask
3 L Erlenmeyer Flask
4 L Erlenmeyer Flask
5 L Erlenmeyer Flask
6 L Erlenmeyer Flask
Unit Size:
Each
Description
- Constructed of stainless steel
- For use with flasks
- Designed to securely hold flask in place
Specifications
Specifications
| Type | Flask Clamp |
| Material | Stainless Steel |
| For Use With (Equipment) | 6 L Erlenmeyer Flask |
| For Use With (Application) | Holding flasks |
| Description | 6 Liter Erlenmeyer Flask Clamp |
| Color | Silver |
| Jaw Type | Four Interlocking |