missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERI1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14592-1-AP-20UL
This item is not returnable.
View return policy
Description
RNA exonuclease that binds to the 3′-end of histone mRNAs and degrades them, suggesting that it plays an essential role in histone mRNA decay after replication. A 2′ and 3′-hydroxyl groups at the last nucleotide of the histone 3′-end is required for efficient degradation of RNA substrates. Also able to degrade the 3′-overhangs of short interfering RNAs (siRNAs) in vitro, suggesting a possible role as regulator of RNA interference (RNAi). Requires for binding the 5′-ACCCA-3′ sequence present in stem-loop structure. Able to bind other mRNAs. Required for 5.8S rRNA 3′-end processing. Also binds to 5.8s ribosomal RNA. Binds with high affinity to the stem-loop structure of replication-dependent histone pre-mRNAs.Specifications
| ERI1 | |
| Polyclonal | |
| Unconjugated | |
| ERI1 | |
| 3 5 exonuclease ERI1, 3 5 exoribonuclease 1, 3EXO, 3′HEXO, Eri 1 homolog, ERI1, exoribonuclease 1, HEXO, Histone mRNA 3 exonuclease 1, Protein 3hExo, THEX1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 361159, 67276, 90459 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5FVR4, Q7TMF2, Q8IV48 | |
| Eri1 | |
| ERI1 Fusion Protein Ag6125 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction