Learn More
ERI1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14592-1-AP-150UL
 View more versions of this product
Product Code. 16813304
  • 410.00 € / 150µL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
RNA exonuclease that binds to the 3′-end of histone mRNAs and degrades them, suggesting that it plays an essential role in histone mRNA decay after replication. A 2′ and 3′-hydroxyl groups at the last nucleotide of the histone 3′-end is required for efficient degradation of RNA substrates. Also able to degrade the 3′-overhangs of short interfering RNAs (siRNAs) in vitro, suggesting a possible role as regulator of RNA interference (RNAi). Requires for binding the 5′-ACCCA-3′ sequence present in stem-loop structure. Able to bind other mRNAs. Required for 5.8S rRNA 3′-end processing. Also binds to 5.8s ribosomal RNA. Binds with high affinity to the stem-loop structure of replication-dependent histone pre-mRNAs.
Specifications
Specifications
ERI1
Polyclonal
Unconjugated
ERI1
3 5 exonuclease ERI1, 3 5 exoribonuclease 1, 3EXO, 3′HEXO, Eri 1 homolog, ERI1, exoribonuclease 1, HEXO, Histone mRNA 3 exonuclease 1, Protein 3hExo, THEX1
Rabbit
Antigen Affinity Chromatography
RUO
361159, 67276, 90459
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5FVR4, Q7TMF2, Q8IV48
Eri1
ERI1 Fusion Protein Ag6125
150 μL
Primary
Human, Mouse, Rat