missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21396-1-AP-150UL
This item is not returnable.
View return policy
Description
ERC2 is a protein coding gene.Specifications
| ERC2 | |
| Polyclonal | |
| Unconjugated | |
| ERC2 | |
| CAST, CAST1, ELKSL, ERC protein 2, ERC2, KIAA0378, SPBC110, Spc110 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 238988, 259269, 26059 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.57 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O15083, Q6PH08, Q8K3M6 | |
| Erc2 | |
| ERC2 Fusion Protein Ag16080 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction