missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ERC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA517181
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody.
The protein encoded by this gene is a member of a family of RIM-binding proteins. RIMs are active zone proteins that regulate neurotransmitter release. This gene has been found fused to the receptor-type tyrosine kinase gene RET by gene rearrangement due to the translocation t(10;12)(q11;p13). Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| ERC1 | |
| Polyclonal | |
| Unconjugated | |
| ERC1 | |
| 4930404L01Rik; 5033405M01Rik; 9630025C19Rik; B430107L16Rik; CAST; CAST2; CAZ-associated structural protein 2; Elks; ELKS/Rab6-interacting/CAST family member 1; ERC; ERC protein 1; Erc1; ERC-1; ERC1b; FLJ31750; KIAA1081; MGC12974; mKIAA1081; OTTHUMP00000160633; Rab6 interacting protein 2; RAB6-interacting protein 2; Rab6ip2; RAB6IP2A; RAB6IP2B; RIM-Binding Protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 111173, 23085, 266806 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 8.11 μg/mL | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
| Q811U3, Q8IUD2, Q99MI1 | |
| ERC1 | |
| Synthetic peptide corresponding to residues near the amino terminus of ERC1 protein. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction