missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ErbB4/Her4 Antibody (182818), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB11311PECY55
This item is not returnable.
View return policy
Description
ErbB4/Her4 Monoclonal antibody specifically detects ErbB4/Her4 in Human samples. It is validated for Flow Cytometry
Specifications
| ErbB4/Her4 | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 182818 | |
| Flow Cytometry | |
| avian erythroblastic leukemia viral (v-erb-b2) oncogene homolog 4, EC 2.7.10, EC 2.7.10.1, HER4MGC138404, p180erbB4, Proto-oncogene-like protein c-ErbB-4, receptor tyrosine-protein kinase erbB-4, Tyrosine kinase-type cell surface receptor HER4, v-erb-a avian erythroblastic leukemia viral oncogene homolog-like 4, v-erb-a erythroblastic leukemia viral oncogene homolog 4 (avian) | |
| Mouse myeloma cell line NS0-derived recombinant human ErbB4/Her4, Glu26-Arg649, Accession # Q15303 | |
| 0.1 mL | |
| Angiogenesis, Cancer, Phospho Specific, Protein Kinase, Tumor Suppressors | |
| 2066 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction