Learn More
Equilin 3-Sulfate-d4 Sodium Salt (stabilized with TRIS, 50% w/w), TRC
Specifications
Specifications
| Chemical Name or Material | Equilin 3-sulfate-d4 sodium salt |
| CAS | 285979-81-9 |
| Molecular Formula | C18 2H4 H15 O5 S . Na |
| Quantity | 1 mg |
| Synonym | Estra-1,3,5(10),7-tetraen-17-one-2,4,16,16-d4, 3-(sulfooxy)-, sodium salt (9CI), Sodium Equilin-2,4,16,16-d4 Sulfate, Equilin 3-Sulfate-d4 Sodium Salt, 3-(Sulfooxy)estra-1,3,5(10),7-tetraen-17-one-2,4,16,16-d4 sodium salt, Equilin sulfate-d4 sodium salt, Equilin Sulfate-2,4,16,16-D4 Sodium Salt, 3-Hydroxyestra-1,3,5(10),7-tetraen-17-one-2,4,16,16-d4 sodium sulfate |
| InChI Formula | InChI=1S/C18H20O5S.Na/c1-18-9-8-14-13-5-3-12(23-24(20,21)22)10-11(13)2-4-15(14)16(18)6-7-17(18)19;/h3-5,10,14,16H,2,6-9H2,1H3,(H,20,21,22);/q;+1/p-1/t14-,16+,18+;/m1./s1/i3D,7D2,10D; |
| SMILES | [Na+].[2H]c1cc2[C@H]3CC[C@@]4(C)[C@@H](CC([2H])([2H])C4=O)C3=CCc2c([2H])c1OS(=O)(=O)[O-] |
| IUPAC Name | sodium;[(9S,13S,14S)-2,4,16,16-tetradeuterio-13-methyl-17-oxo-6,9,11,12,14,15-hexahydrocyclopenta[a]phenanthren-3-yl] sulfate |
| Molecular Weight (g/mol) | 374.42 |
| Formula Weight | 374.1102 |